KEY007387395 Key Organics / BIONET

(2E)-3-(4-benzoyl-1-methyl-1H-pyrrol-2-yl)prop-2-enoic acid

  • MDL number MFCD03791143
  • Cas Number : 128843-46-9
  • Molecular Weight : 255.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 180
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3146

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0299_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart