KEY007387398 Key Organics / BIONET

3-methyl-4-[3-(trifluoromethyl)phenyl]-1,2-oxazol-5-amine

  • MDL number MFCD03791146
  • Cas Number : 446276-00-2
  • Molecular Weight : 242.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 123
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3193

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0381_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart