KEY007387406 Key Organics / BIONET

5-chloro-1-{[3-(trifluoromethyl)phenyl]methyl}-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00141725
  • Cas Number : 89159-26-2
  • Molecular Weight : 339.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165.5 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5393

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11L-353S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart