KEY007387408 Key Organics / BIONET

2-{[3-(trifluoromethyl)phenyl]methanimidamido}acetic acid

  • MDL number MFCD01935986
  • Cas Number : 338982-13-1
  • Molecular Weight : 246.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5728

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11M-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart