KEY007387412 Key Organics / BIONET

3-methyl-1-(4-nitrophenyl)-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD03791150
  • Cas Number : 852690-94-9
  • Molecular Weight : 362.312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 165
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6875

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0223_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart