KEY007387413 Key Organics / BIONET

ethyl 7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD03791151
  • Cas Number : 852690-95-0
  • Molecular Weight : 365.312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273 - 275
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6888

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0242_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart