KEY007387416 Key Organics / BIONET

1,3-dimethyl-1H,6H-[1,2]diazolo[3,4-c]pyrazole

  • MDL number MFCD00238783
  • Cas Number : 446276-19-3
  • Molecular Weight : 136.158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218 - 220
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6910

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0266_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart