KEY007387421 Key Organics / BIONET

2-chloro-5-(2,3-dihydro-1H-isoindol-2-yl)-4-fluorophenol

  • MDL number MFCD03791152
  • Cas Number : 852690-96-1
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137 - 139
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6920

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0289_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart