KEY007387422 Key Organics / BIONET

(E)-2-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-N'-hydroxyethanimidamide

  • MDL number MFCD03791153
  • Cas Number : 656223-02-8
  • Molecular Weight : 267.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 - 152
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6922

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart