KEY007387426 Key Organics / BIONET

[3-(pyrimidin-2-yloxy)phenyl]methanol

  • MDL number MFCD03791155
  • Cas Number : 82440-25-3
  • Molecular Weight : 202.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 150
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6936

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0316_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart