KEY007387430 Key Organics / BIONET

(Z)-N'-hydroxy-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzene-1-carboximidamide

  • MDL number MFCD00663373
  • Cas Number : 263161-34-8
  • Molecular Weight : 297.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 132 - 133
Iupac Name N'-hydroxy-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarboximidamide
Inchi Code 1S/C13H10F3N3O2/c14-13(15,16)9-3-6-11(18-7-9)21-10-4-1-8(2-5-10)12(17)19-20/h1-7,12H,17H2
Inchi Key VVLZLUQBKVMYQQ-UHFFFAOYSA-N

Description

Melting Point 132 - 133
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6942

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0329_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart