KEY007387434 Key Organics / BIONET

2-[(E)-2-(furan-3-yl)ethenyl]-5-hydroxy-1-benzofuran-3-carbaldehyde

  • MDL number MFCD03791159
  • Cas Number : 852691-02-2
  • Molecular Weight : 254.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225 - 228
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6952

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0343_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart