KEY007387435 Key Organics / BIONET

5-[(E)-2-phenylethenyl]-2H-1,2,3,4-tetrazole

  • MDL number MFCD00276928
  • Cas Number : 220429-71-0
  • Molecular Weight : 172.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 187
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7051

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0832_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart