KEY007387437 Key Organics / BIONET

4-[(2-chloro-6-fluorophenyl)methyl]-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD00120327
  • Cas Number : 387352-96-7
  • Molecular Weight : 238.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7062

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart