KEY007387438 Key Organics / BIONET

3-{[1,1'-biphenyl]-4-yl}-1H-pyrazole

  • MDL number MFCD02755978
  • Cas Number : 446276-22-8
  • Molecular Weight : 220.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 161
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7103

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0899_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart