KEY007387442 Key Organics / BIONET

5-(4-methylpiperazin-1-yl)-2-nitroaniline

  • MDL number MFCD00545964
  • Cas Number : 23491-48-7
  • Molecular Weight : 236.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152 - 155
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9608

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0229_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart