KEY007387445 Key Organics / BIONET

3-methyl-4-phenyl-1H-pyrazole

  • MDL number MFCD00051752
  • Cas Number : 13788-84-6
  • Molecular Weight : 158.204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 122
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9758

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0222_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart