KEY007387447 Key Organics / BIONET

5-hydroxy-2-[(E)-2-(3-methoxyphenyl)ethenyl]naphtho[1,2-b]furan-3-carbaldehyde

  • MDL number MFCD03791162
  • Cas Number : 853097-49-1
  • Molecular Weight : 344.366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232 - 234
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9819

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart