KEY007387449 Key Organics / BIONET

(Z)-N'-hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide

  • MDL number MFCD02090466
  • Cas Number : 690632-15-6
  • Molecular Weight : 234.178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 78
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9832

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0313_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart