KEY007387452 Key Organics / BIONET

3-amino-1-(2-chlorophenyl)urea

  • MDL number MFCD03791165
  • Cas Number : 35580-76-8
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 114 - 116
Iupac Name N-(2-chlorophenyl)hydrazinecarboxamide
Inchi Code 1S/C7H8ClN3O/c8-5-3-1-2-4-6(5)10-7(12)11-9/h1-4H,9H2,(H2,10,11,12)
Inchi Key GCGJAIHYYAKDOJ-UHFFFAOYSA-N

Description

Melting Point 114 - 116
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9913

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12T-0883_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart