KEY007387453 Key Organics / BIONET

2-methyl-6-nitro-1,3-benzoxazole

  • MDL number MFCD00022848
  • Cas Number : 5683-43-2
  • Molecular Weight : 178.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9914

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0885_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart