KEY007387454 Key Organics / BIONET

3-amino-1-(2,4-dimethoxyphenyl)urea

  • MDL number MFCD03791166
  • Cas Number : 853097-58-2
  • Molecular Weight : 211.221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141 - 143
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9930

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart