KEY007387459 Key Organics / BIONET

2-[(3-nitropyridin-2-yl)sulfanyl]aniline

  • MDL number MFCD01936009
  • Cas Number : 92316-06-8
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125.5 - 127.5
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Red Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63785

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-339S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart