KEY007387460 Key Organics / BIONET

4-(1H-imidazol-1-yl)-3-nitrobenzoic acid

  • MDL number MFCD01935984
  • Cas Number : 167626-67-7
  • Molecular Weight : 233.183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 267 - 269
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63873

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1G-449S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart