KEY007387463 Key Organics / BIONET

N-(4-methoxy-2-nitrophenyl)acetamide

  • MDL number MFCD00017018
  • Cas Number : 119-81-3
  • Molecular Weight : 210.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 118
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65011

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart