KEY007387475 Key Organics / BIONET

2-{3-[(1E)-2-nitroprop-1-en-1-yl]phenoxy}pyrimidine

  • MDL number MFCD03791172
  • Cas Number : 855308-69-9
  • Molecular Weight : 257.249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81 - 83
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0251_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart