KEY007387477 Key Organics / BIONET

5-nitro-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD01159700
  • Cas Number : 6345-55-7
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239 - 241
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66053

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0255_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart