KEY007387480 Key Organics / BIONET

4-(pyrimidin-2-yloxy)benzaldehyde

  • MDL number MFCD03791174
  • Cas Number : 433920-92-4
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 76 - 78
Iupac Name 4-(2-pyrimidinyloxy)benzaldehyde
Inchi Code 1S/C11H8N2O2/c14-8-9-2-4-10(5-3-9)15-11-12-6-1-7-13-11/h1-8H
Inchi Key CGSRJSIAGKSMHU-UHFFFAOYSA-N

Description

Melting Point 76 - 78
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66057

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0265_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart