KEY007387481 Key Organics / BIONET

1-(6,7-dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)ethan-1-one

  • MDL number MFCD03791175
  • Cas Number : 166816-11-1
  • Molecular Weight : 247.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 89
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66064

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart