KEY007387488 Key Organics / BIONET

methyl 2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate

  • MDL number MFCD00662517
  • Cas Number : 259654-73-4
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111 - 113
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66137

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0674_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart