KEY007387499 Key Organics / BIONET

5-(bromomethyl)-1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole

  • MDL number MFCD01935938
  • Cas Number : 338982-24-4
  • Molecular Weight : 299.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 100
Purity 95
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68236

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2M-331S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart