KEY007387504 Key Organics / BIONET

[(2,4-dichlorophenyl)amino]thiourea

  • MDL number MFCD03791186
  • Cas Number : 13124-11-3
  • Molecular Weight : 236.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68584

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart