KEY007387506 Key Organics / BIONET

[(4-chlorophenyl)amino]thiourea

  • MDL number MFCD00456275
  • Cas Number : 7382-41-4
  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68587

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart