KEY007387510 Key Organics / BIONET

5-nitro-2-(pyrrolidin-1-yl)pyridine

  • MDL number MFCD00119573
  • Cas Number : 26820-63-3
  • Molecular Weight : 193.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137 - 139
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69416

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0268_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart