KEY007387514 Key Organics / BIONET

ethyl 4-hydroxy-5-oxo-1-[(thiophen-2-yl)methyl]-2,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD03791192
  • Cas Number : 131436-78-7
  • Molecular Weight : 267.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 98 - 100
Iupac Name ethyl 4-hydroxy-5-oxo-1-(2-thienylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate
Inchi Code 1S/C12H13NO4S/c1-2-17-12(16)9-7-13(11(15)10(9)14)6-8-4-3-5-18-8/h3-5,9H,2,6-7H2,1H3
Inchi Key RFZBVKYVEUTCMN-UHFFFAOYSA-N

Description

Melting Point 98 - 100
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69675

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0263_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart