KEY007387516 Key Organics / BIONET

7-amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD03791193
  • Cas Number : 863305-82-2
  • Molecular Weight : 336.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224 - 226
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69682

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0276_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart