KEY007387517 Key Organics / BIONET

5-(morpholin-4-yl)-2-nitrobenzamide

  • MDL number MFCD03791194
  • Cas Number : 404009-38-7
  • Molecular Weight : 251.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280 - 282
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69693

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0291_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart