KEY007387527 Key Organics / BIONET

methyl 4-(4-chlorophenyl)-2,4-dioxobutanoate

  • MDL number MFCD03001315
  • Cas Number : 39757-35-2
  • Molecular Weight : 240.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 118 - 120
Iupac Name methyl (2E)-4-(4-chlorophenyl)-2-hydroxy-4-oxo-2-butenoate
Inchi Code 1S/C11H9ClO4/c1-16-11(15)10(14)6-9(13)7-2-4-8(12)5-3-7/h2-5H,6H2,1H3
Inchi Key PIULUHUQDVELBO-UHFFFAOYSA-N

Description

Melting Point 118 - 120
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-606S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart