KEY007387529 Key Organics / BIONET

5-amino-3-methyl-1,2-oxazole-4-carbonitrile

  • MDL number MFCD00027378
  • Cas Number : 35261-01-9
  • Molecular Weight : 123.115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227 - 230
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-382S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart