KEY007387530 Key Organics / BIONET

methyl 4-(4-formyl-2-nitrophenoxy)-3-methoxybenzoate

  • MDL number MFCD03425757
  • Cas Number : 439095-22-4
  • Molecular Weight : 331.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 118
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73085

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0206_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart