KEY007387536 Key Organics / BIONET

5-(morpholin-4-yl)-2-nitroaniline

  • MDL number MFCD00511844
  • Cas Number : 54998-00-4
  • Molecular Weight : 223.232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182 - 185
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73170

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0352_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart