KEY007387539 Key Organics / BIONET

4-(3-bromothiophen-2-yl)-6-methylpyrimidin-2-amine

  • MDL number MFCD03791203
  • Cas Number : 886360-55-0
  • Molecular Weight : 270.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 153 - 154
Iupac Name 4-(3-bromo-2-thienyl)-6-methyl-2-pyrimidinamine
Inchi Code 1S/C9H8BrN3S/c1-5-4-7(13-9(11)12-5)8-6(10)2-3-14-8/h2-4H,1H3,(H2,11,12,13)
Inchi Key KUTYUJWSFOAFHN-UHFFFAOYSA-N

Description

Melting Point 153 - 154
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73186

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0610_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart