KEY007387540 Key Organics / BIONET

3-[4-(methoxycarbonyl)phenyl]-3-(2,2,2-trifluoroacetamido)propanoic acid

  • MDL number MFCD03791204
  • Cas Number : 886360-56-1
  • Molecular Weight : 319.236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 173
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0625_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart