KEY007387542 Key Organics / BIONET

methyl 4-[2-carbamoyl-1-(2,2,2-trifluoroacetamido)ethyl]benzoate

  • MDL number MFCD03791206
  • Cas Number : 886360-57-2
  • Molecular Weight : 318.252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231 - 233
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0646_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart