KEY007387543 Key Organics / BIONET

2-(3-nitrophenyl)quinoxaline

  • MDL number MFCD00430477
  • Cas Number : 5021-44-3
  • Molecular Weight : 251.245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 185
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73204

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0651_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart