KEY007387544 Key Organics / BIONET

[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanamine

  • MDL number MFCD03791207
  • Cas Number : 886360-58-3
  • Molecular Weight : 238.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73362

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0217_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart