KEY007387545 Key Organics / BIONET

1-[3-(trifluoromethyl)phenyl]-1H-pyrazol-4-ol

  • MDL number MFCD03791208
  • Cas Number : 77458-38-9
  • Molecular Weight : 228.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73364

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart