KEY007387547 Key Organics / BIONET

methyl 4-hydroxy-5-oxo-1-[(pyridin-2-yl)methyl]-2,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD03791209
  • Cas Number : 885951-47-3
  • Molecular Weight : 248.238

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 149
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73371

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0231_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart