KEY007387548 Key Organics / BIONET

methyl 5-chloro-6-(phenylsulfanyl)pyridine-3-carboxylate

  • MDL number MFCD03791210
  • Cas Number : 885951-48-4
  • Molecular Weight : 279.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 103
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73375

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0238_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart