KEY007387552 Key Organics / BIONET

3-(4-chlorophenyl)-3-(2,2,2-trifluoroacetamido)propanamide

  • MDL number MFCD03012828
  • Cas Number : 439110-82-4
  • Molecular Weight : 294.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 241 - 243
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73392

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0273_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart